APO455836857 Apollo Scientific Ltd

2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone

  • MDL number MFCD13173364
  • Cas Number : 489429-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methoxy-6-(trifluoroacetyl)aniline, 2-Amino-3-(trifluoroacetyl)anisole, 1-(2-Amino-3-methoxyphenyl)-2,2,2-trifluoroethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48286

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48286
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart