APO455836858 Apollo Scientific Ltd

2-(Bromomethyl)-4-fluorobenzaldehyde

  • MDL number MFCD18205513
  • Cas Number : 1379346-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Fluoro-2-formylbenzyl bromide, alpha-Bromo-4-fluoro-o-tolualdehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64023

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=64023
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart