APO455836860 Apollo Scientific Ltd

5-(Bromomethyl)-2-fluorobenzaldehyde

  • MDL number MFCD28096626
  • Cas Number : 1820717-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-3-formylbenzyl bromide, alpha-Bromo-6-fluoro-m-tolualdehyde
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64054

Safety Information

signalword Danger
pcodes P260;P280;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64054
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart