APO455836862 Apollo Scientific Ltd

3'-Bromo-2,2,2,2'-tetrafluoroacetophenone

  • MDL number MFCD25541891
  • Cas Number : 871353-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3-Bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-one
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54316

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54316
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart