APO455836870 Apollo Scientific Ltd

2,6-Difluoro-3-nitropyridine

  • MDL number MFCD11044323
  • Cas Number : 58602-02-1
  • Molecular Weight : 160.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-nitropyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-nitropyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart