APO455836872 Apollo Scientific Ltd

2-Fluoro-5-nitrophenol

  • MDL number MFCD09054718
  • Cas Number : 22510-08-3
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-5-nitrophenol
Inchi Code 1S/C6H4FNO3/c7-5-2-1-4(8(10)11)3-6(5)9/h1-3,9H
Inchi Key WFRLFZAMCVAQLN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-nitrophenol
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart