APO455836875 Apollo Scientific Ltd

3-(Pentafluorothio)benzyl bromide

  • MDL number MFCD16652411
  • Cas Number : 1056893-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Bromomethyl)phenylsulphur pentafluoride, 1-(Bromomethyl)-3-(pentafluorosulphanyl)benzene
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54714

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54714
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart