APO455836880 Apollo Scientific Ltd

4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD06801688
  • Cas Number : 874219-35-9
  • Molecular Weight : 197.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-255
Synonym Methyl 5-borono-2-fluorobenzoate
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart