APO455836882 Apollo Scientific Ltd

1-Bromo-1,1,2,2-tetrafluorohexane

  • MDL number MFCD25541897
  • Cas Number : 151831-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 122-124
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54711

Safety Information

signalword Warning
pcodes P280;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54711
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart