APO455836884 Apollo Scientific Ltd

5-Fluoro-2-iodobenzaldehyde

  • MDL number MFCD07782057
  • Cas Number : 877264-44-3
  • Molecular Weight : 250.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-2-iodobenzaldehyde
Inchi Code 1S/C7H4FIO/c8-6-1-2-7(9)5(3-6)4-10/h1-4H
Inchi Key KSDLYLRIDDWCNS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-iodobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart