APO455836894 Apollo Scientific Ltd

2-Chloro-3-fluoropyridine-4-boronic acid

  • MDL number MFCD04972403
  • Cas Number : 937595-71-6
  • Molecular Weight : 175.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-3-fluoro-4-pyridinylboronic acid
Inchi Code 1S/C5H4BClFNO2/c7-5-4(8)3(6(10)11)1-2-9-5/h1-2,10-11H
Inchi Key ZXGCWGYBIXCFMP-UHFFFAOYSA-N

Description

Synonym 4-Borono-2-chloro-3-fluoropyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-fluoropyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-fluoropyridine-4-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart