APO455836901 Apollo Scientific Ltd

2,3,5-Trifluorobenzenesulphonyl chloride

  • MDL number MFCD08532462
  • Cas Number : 914636-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-50
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10904

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10904
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart