APO455836907 Apollo Scientific Ltd

2-Methoxy-5-(trifluoromethyl)pyridine

  • MDL number MFCD00153203
  • Cas Number : 175277-45-9
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-5-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart