APO455836911 Apollo Scientific Ltd

1-Methyl-2-(trifluoromethyl)benzene

  • MDL number MFCD00153204
  • Cas Number : 13630-19-8
  • Molecular Weight : 160.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha,alpha,alpha-Trifluoro-o-xylene
Purity 97
Density 1.17
Flash Point 26
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 125-126
Product Link https://store.apolloscientific.co.uk/product/1-methyl-2-trifluoromethylbenzene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-2-trifluoromethylbenzene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart