APO455836913 Apollo Scientific Ltd

Ethyl difluoro(phenyl)acetate

  • MDL number MFCD08275296
  • Cas Number : 2248-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Difluoro(phenyl)acetic acid ethyl ester, Ethyl 2,2-difluoro-2-phenylethanoate
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70-72/1mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-difluorophenylacetate-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-difluorophenylacetate-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart