APO455836915 Apollo Scientific Ltd

Methyl 2-chloro-4-fluorobenzoate

  • MDL number MFCD00173937
  • Cas Number : 85953-29-3
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 103-105/15mm
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloro-4-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloro-4-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart