APO455836918 Apollo Scientific Ltd

Methyl 3,5-bis(trifluoromethyl)benzoate

  • MDL number MFCD00052317
  • Cas Number : 26107-80-2
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85-87/15mm
Product Link https://store.apolloscientific.co.uk/product/methyl-35-bistrifluoromethylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-35-bistrifluoromethylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart