APO455836919 Apollo Scientific Ltd

3-Fluoro-5-methoxybenzonitrile

  • MDL number MFCD06797883
  • Cas Number : 439280-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-100
Synonym 3-Cyano-5-fluoroanisole, 5-Fluoro-m-anisonitrile
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart