APO455836920 Apollo Scientific Ltd

2-Bromo-3-fluoroaniline

  • MDL number MFCD07369915
  • Cas Number : 111721-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroaniline-98

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroaniline-98
pictograms GHS07
hstatements H312;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart