APO455836921 Apollo Scientific Ltd

2-Bromo-3-fluoronitrobenzene

  • MDL number MFCD07782080
  • Cas Number : 59255-94-6
  • Molecular Weight : 220

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-45
Synonym 2-Bromo-1-fluoro-3-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart