APO455836930 Apollo Scientific Ltd

2-Bromo-5-iodobenzotrifluoride

  • MDL number MFCD11855999
  • Cas Number : 364-11-4
  • Molecular Weight : 350.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Synonym 1-Bromo-4-iodo-2-(trifluoromethyl)benzene, 2-Bromo-5-iodo-alpha,alpha,alpha-trifluorotoluene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-iodobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-iodobenzotrifluoride
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart