APO455836931 Apollo Scientific Ltd

3,5-Dibromobenzotrifluoride

  • MDL number MFCD00526219
  • Cas Number : 401-84-3
  • Molecular Weight : 303.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,3-Dibromo-5-(trifluoromethyl)benzene, 3,5-Dibromo-alpha,alpha,alpha-trifluorotoluene
Purity 96
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dibromobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dibromobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart