APO455836933 Apollo Scientific Ltd

2,2,2,3',5'-Pentafluoroacetophenone

  • MDL number MFCD01319993
  • Cas Number : 845823-12-3
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3,5-Difluorophenyl)-2,2,2-trifluoroethan-1-one
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22235-pentafluoroacetophenone

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/22235-pentafluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart