APO455836936 Apollo Scientific Ltd

5-Fluoro-1-methyl-1H-indazole-6-boronic acid

  • MDL number MFCD18089818
  • Cas Number : 2096331-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Borono-5-fluoro-1-methyl-1H-indazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48394

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48394
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart