APO455836937 Apollo Scientific Ltd

6-Bromo-5-fluoro-2-methyl-2H-indazole

  • MDL number MFCD18089820
  • Cas Number : 1286734-87-9
  • Molecular Weight : 229.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-5-fluoro-2-methyl-2h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-5-fluoro-2-methyl-2h-indazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart