APO455836939 Apollo Scientific Ltd

6-Bromo-5-fluoro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

  • MDL number MFCD18384869
  • Cas Number : 1286734-76-6
  • Molecular Weight : 299.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(6-Bromo-5-fluoro-1H-indazol-1-yl)tetrahydro-2H-pyran, 2-(6-Bromo-5-fluoro-1H-indazol-1-yl)oxane
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-5-fluoro-1-tetrahydro-2h-pyran-2-yl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-5-fluoro-1-tetrahydro-2h-pyran-2-yl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart