APO455836940 Apollo Scientific Ltd

5-Fluoro-1-methyl-1H-indazole-6-carboxaldehyde

  • MDL number MFCD18384870
  • Cas Number : 1286734-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Fluoro-6-formyl-1-methyl-1H-indazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48403

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48403
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart