APO455836952 Apollo Scientific Ltd

2,5-Dichloro-3-(difluoromethyl)pyridine

  • MDL number MFCD18384828
  • Cas Number : 71701-93-4
  • Molecular Weight : 198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,5-Dichloro-alpha,alpha-difluoro-3-picoline
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-dichloro-3-difluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/25-dichloro-3-difluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart