APO455836956 Apollo Scientific Ltd

Methyl 4-fluoro-3-iodobenzoate

  • MDL number MFCD11870177
  • Cas Number : 1121586-29-5
  • Molecular Weight : 280.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Purity 97
Density 1.82
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 287-288
Product Link https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-iodobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-fluoro-3-iodobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart