APO455836959 Apollo Scientific Ltd

4-Fluoro-3-iodonitrobenzene

  • MDL number MFCD08461002
  • Cas Number : 177363-10-9
  • Molecular Weight : 267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-2-iodo-4-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-iodonitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-iodonitrobenzene
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart