APO455836961 Apollo Scientific Ltd

2,4-Difluoro-5-iodonitrobenzene

  • MDL number MFCD20322039
  • Cas Number : 148388-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,5-Difluoro-2-iodo-4-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 65-67/0.1mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49828

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49828
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart