APO455836963 Apollo Scientific Ltd

2,6-Difluoro-3-hydroxypyridine

  • MDL number MFCD11044351
  • Cas Number : 209328-85-8
  • Molecular Weight : 131.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-125
Synonym 2,6-Difluoropyridin-3-ol
Purity 96
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-hydroxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-hydroxypyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart