APO455836965 Apollo Scientific Ltd

2-Bromo-5-fluoro-4-hydroxypyridine

  • MDL number MFCD13189884
  • Cas Number : 1196152-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-fluoropyridin-4-ol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-hydroxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart