APO455836966 Apollo Scientific Ltd

2-Chloro-3-fluoro-4-hydroxypyridine

  • MDL number MFCD13189885
  • Cas Number : 1184172-46-0
  • Molecular Weight : 147.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-3-fluoropyridin-4-ol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-fluoro-4-hydroxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-fluoro-4-hydroxypyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart