APO455836967 Apollo Scientific Ltd

3-Fluoro-5-methoxybenzoic acid

  • MDL number MFCD08235092
  • Cas Number : 176548-72-4
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-133
Synonym 5-Fluoro-m-anisic acid, 3-Carboxy-5-fluoroanisole
Purity 98
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart