APO455836969 Apollo Scientific Ltd

2-Fluoro-5-hydroxy-3-methylpyridine

  • MDL number MFCD09839279
  • Cas Number : 186593-50-0
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Synonym 6-Fluoro-5-methylpyridin-3-ol, 2-Fluoro-5-hydroxy-3-picoline
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxy-3-methylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-hydroxy-3-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart