APO455836971 Apollo Scientific Ltd

2-Fluoro-3-hydroxy-5-methylpyridine

  • MDL number MFCD16611070
  • Cas Number : 1184172-53-9
  • Molecular Weight : 127.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-5-methylpyridin-3-ol, 6-Fluoro-5-hydroxy-3-picoline
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-hydroxy-5-methylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-hydroxy-5-methylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart