APO455836975 Apollo Scientific Ltd

2-Bromo-5-fluoro-4-nitrotoluene

  • MDL number MFCD11110550
  • Cas Number : 170098-98-3
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-4-fluoro-2-methyl-5-nitrobenzene, 5-Bromo-2-fluoro-4-methylnitrobenzene
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-fluoro-4-nitrotoluene
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart