APO455836976 Apollo Scientific Ltd

2-Amino-5-bromo-4-fluorobenzoic acid

  • MDL number MFCD06409112
  • Cas Number : 143945-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-4-fluoroanthranilic acid, 4-Bromo-2-carboxy-5-fluoroaniline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-fluorobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart