APO455836979 Apollo Scientific Ltd

2-Fluoro-6-methylbenzyl alcohol

  • MDL number MFCD06660280
  • Cas Number : 478163-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Fluoro-6-methylphenyl)methanol, (6-Fluoro-o-tolyl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-methylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart