APO455836983 Apollo Scientific Ltd

2-Methyl-3-nitrobenzotrifluoride

  • MDL number MFCD00042322
  • Cas Number : 6656-49-1
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Methyl-3-(trifluoromethyl)nitrobenzene, 2-Methyl-1-nitro-3-(trifluoromethyl)benzene
Purity 97
Flash Point >100
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86/10mm
Product Link https://store.apolloscientific.co.uk/product/2-methyl-3-nitrobenzotrifluoride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-3-nitrobenzotrifluoride
pictograms GHS06
hstatements H301+H311;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart