APO455836984 Apollo Scientific Ltd

3-Bromo-4-hydroxyphenylsulphur pentafluoride

  • MDL number MFCD23166763
  • Cas Number : 1426290-12-1
  • Molecular Weight : 299.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-4-(pentafluorosulphanyl)phenol, 2-Bromo-4-(pentafluorothio)phenol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-hydroxyphenylsulphur-pentafluoride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-hydroxyphenylsulphur-pentafluoride
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart