APO455836995 Apollo Scientific Ltd

2-Amino-4-[(trifluoromethyl)thio]phenol

  • MDL number MFCD23703635
  • Cas Number : 228401-48-7
  • Molecular Weight : 209.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-4-[(trifluoromethyl)sulphanyl]phenol, 2-Hydroxy-5-[(trifluoromethyl)thio]aniline
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethylthiophenol_PC51173

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-trifluoromethylthiophenol_PC51173
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart