APO455836997 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethoxy)phenacyl bromide

  • MDL number MFCD23703637
  • Cas Number : 1429321-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-[2-fluoro-3-(trifluoromethoxy)phenyl]ethan-1-one, 2-Bromo-2'-fluoro-3'-(trifluoromethoxy)acetophenone
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51978

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51978
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart