APO455837001 Apollo Scientific Ltd

2,4-Difluoro-3-(trifluoromethyl)benzaldehyde

  • MDL number MFCD12404958
  • Cas Number : 134099-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha,alpha,alpha,2,4-Pentafluoro-m-tolualdehyde, 2,6-Difluoro-3-formylbenzotrifluoride
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52143

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52143
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart