APO455837004 Apollo Scientific Ltd

5-Bromo-3-fluoro-2-hydroxybenzoic acid

  • MDL number MFCD06203615
  • Cas Number : 251300-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-fluorosalicylic acid, 4-Bromo-2-carboxy-6-fluorophenol
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52290

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52290
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart