APO455837006 Apollo Scientific Ltd

3'-(Pentafluorothio)acetophenone

  • MDL number MFCD16652407
  • Cas Number : 159896-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3-(Pentafluorosulphanyl)phenyl]ethan-1-one, 3-Acetylphenylsulphur pentafluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52975

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52975
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart