APO455837008 Apollo Scientific Ltd

4-Bromo-6-(pentafluorothio)-1H-benzimidazole

  • MDL number MFCD24369706
  • Cas Number : 1432075-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Bromo-1H-benzimidazol-6-yl)sulphur pentafluoride, 4-Bromo-6-(pentafluorosulphanyl)-1H-benzimidazole
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52984

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52984
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart