APO455837010 Apollo Scientific Ltd

2-Bromo-6-methylphenyl trifluoromethanesulphonate

  • MDL number MFCD27975433
  • Cas Number : 1820716-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-6-methylphenyl triflate, 3-Bromo-2-{[(trifluoromethyl)sulphonyl]oxy}toluene
Purity 95
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61626

Safety Information

signalword Danger
pcodes P280;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61626
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart