APO455837013 Apollo Scientific Ltd

2-(Difluoromethyl)-4-hydroxypyridine

  • MDL number MFCD18820876
  • Cas Number : 501125-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Difluoromethyl)pyridin-4-ol, alpha,alpha-Difluoro-4-hydroxy-2-picoline
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52276

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52276
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart